siRNA Details
| Gene Symbol: Crybb3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_031690.1 | NP_113878.1 | 2 | 214 | ||
| XM_006249572.1 | XP_006249634.1 | 3 | 252 | |||
| GenBank mRNA | AF287304.1 | - | 2 | 186 | ||
| X05899.1 | - | 2 | 214 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Crybb3| Gene Symbol: Crybb3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_031690.1 | NP_113878.1 | 2 | 214 | ||
| XM_006249572.1 | XP_006249634.1 | 3 | 252 | |||
| GenBank mRNA | AF287304.1 | - | 2 | 186 | ||
| X05899.1 | - | 2 | 214 | |||