siRNA Details
Gene Symbol: Dpys | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031705.1 | NP_113893.1 | 4 | 870 | ||
GenBank mRNA | BC081768.1 | - | 4 | 825 | 7107549 | |
D63704.1 | - | 4 | 870 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Dpys | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031705.1 | NP_113893.1 | 4 | 870 | ||
GenBank mRNA | BC081768.1 | - | 4 | 825 | 7107549 | |
D63704.1 | - | 4 | 870 |