siRNA Details
| Gene Symbol: Sv2c | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_031593.1 | NP_113781.1 | 6, 7 | 1359 | ||
| GenBank mRNA | AF060174.1 | - | 6, 7 | 1359 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Sv2c| Gene Symbol: Sv2c | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_031593.1 | NP_113781.1 | 6, 7 | 1359 | ||
| GenBank mRNA | AF060174.1 | - | 6, 7 | 1359 | ||