siRNA Details
| Gene Symbol: Ywhae | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_031603.1 | NP_113791.1 | 6 | 842 | ||
| GenBank mRNA | BC063163.1 | - | 6 | 820 | 6921685 | |
| D30739.1 | - | 6 | 826 | |||
| FQ215270.1 | - | 4 | 500 | |||
| FQ220393.1 | - | 6 | 847 | |||
| M84416.1 | - | 6 | 822 | |||
| U53882.1 | - | 6 | 842 | |||

Ywhae
Genomic Map