siRNA Details
| Gene Symbol: Cyp4a8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_031605.2 | NP_113793.2 | 7 | 862 | ||
| GenBank mRNA | BC081771.1 | - | 7 | 862 | 7108258 | |
| M37828.1 | - | 7 | 852 | |||
| Gene Symbol: LOC103692442 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_008763954.1 | XP_008762176.1 | 3 | 1092 | ||

Cyp4a8
Genomic Map