siRNA Details
| Gene Symbol: Pten | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_031606.1 | NP_113794.1 | 2 | 148 | ||
| GenBank mRNA | AF017185.1 | - | 2 | 148 | ||
| AF455569.1 | - | 2 | 148 | |||
More Information
Other Species Targeted
This siRNA also targets the following transcripts in Mouse: NM_008960, U92437, AK088717, BC021445, AK076980, and others.

Pten
Genomic Map