siRNA Details
| Gene Symbol: Klf6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_031642.4 | NP_113830.2 | 2 | 875 | ||
| XM_006254182.1 | XP_006254244.1 | 9 | 2141 | |||
| GenBank mRNA | AF001417.1 | - | 2 | 655 | ||
| AY318957.1 | - | 9 | 2140 | |||
| BC097306.1 | - | 2 | 800 | 7444817 | ||
| FQ225029.1 | - | 1 | 431 | |||
| FQ230654.1 | - | 1 | 525 | |||
| FQ231202.1 | - | 1 | 215 | |||
| FQ231451.1 | - | 1 | 215 | |||
| FQ232231.1 | - | 1 | 214 | |||

Klf6
Genomic Map