siRNA Details
Gene Symbol: Slc15a2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031672.2 | NP_113860.2 | 10 | 1082 | ||
GenBank mRNA | BC085327.1 | - | 10 | 1082 | 7098404 | |
D63149.1 | - | 10 | 1070 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Slc15a2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031672.2 | NP_113860.2 | 10 | 1082 | ||
GenBank mRNA | BC085327.1 | - | 10 | 1082 | 7098404 | |
D63149.1 | - | 10 | 1070 |