siRNA Details
| Gene Symbol: St3gal2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_031695.2 | NP_113883.2 | 4, 5 | 835 | ||
| XM_006255625.2 | XP_006255687.1 | 6, 7 | 2010 | |||
| XM_006255626.2 | XP_006255688.1 | 7, 8 | 2202 | |||
| XM_006255627.2 | XP_006255689.1 | 5, 6 | 1892 | |||
| GenBank mRNA | CO404237.1 | - | 2, 3 | 225 | ||
| X76988.1 | - | 847 | ||||

St3gal2
Genomic Map