siRNA Details
| Gene Symbol: Rpn2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_031698.1 | NP_113886.1 | 5 | 550 | ||
| XM_006235379.1 | XP_006235441.1 | 5 | 582 | |||
| GenBank mRNA | BC060556.1 | - | 6 | 560 | 6919638 | |
| X55298.1 | - | 5 | 550 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Rpn2| Gene Symbol: Rpn2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_031698.1 | NP_113886.1 | 5 | 550 | ||
| XM_006235379.1 | XP_006235441.1 | 5 | 582 | |||
| GenBank mRNA | BC060556.1 | - | 6 | 560 | 6919638 | |
| X55298.1 | - | 5 | 550 | |||