siRNA Details
| Gene Symbol: Arl2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_031711.1 | NP_113899.1 | 3 | 271 | ||
| XM_006230899.1 | XP_006230961.1 | 3 | 320 | |||
| GenBank mRNA | FQ213929.1 | - | 3 | 279 | ||
| Y12708.1 | - | 3 | 271 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Arl2| Gene Symbol: Arl2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_031711.1 | NP_113899.1 | 3 | 271 | ||
| XM_006230899.1 | XP_006230961.1 | 3 | 320 | |||
| GenBank mRNA | FQ213929.1 | - | 3 | 279 | ||
| Y12708.1 | - | 3 | 271 | |||