siRNA Details
| Gene Symbol: Clns1a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_031719.1 | NP_113907.1 | 2 | 251 | ||
| XM_006229792.2 | XP_006229854.1 | 2 | 314 | |||
| GenBank mRNA | BC060555.1 | - | 2 | 295 | 6918578 | |
| D13985.1 | - | 2 | 321 | |||
| D17698.1 | - | 2 | 234 | |||
| L26450.1 | - | 2 | 251 | |||

Clns1a
Genomic Map