siRNA Details
| Gene Symbol: Kcnd2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_031730.2 | NP_113918.2 | 1 | 656 | ||
| GenBank mRNA | M59980.1 | - | 1 | 335 | ||
| S64320.1 | - | 1 | 654 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Kcnd2| Gene Symbol: Kcnd2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_031730.2 | NP_113918.2 | 1 | 656 | ||
| GenBank mRNA | M59980.1 | - | 1 | 335 | ||
| S64320.1 | - | 1 | 654 | |||