siRNA Details
| Gene Symbol: Kcnd3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001270962.1 | NP_001257891.1 | 2 | 313 | ||
| NM_001270963.1 | NP_001257892.1 | 2 | 313 | |||
| NM_031739.3 | NP_113927.2 | 2 | 313 | |||
| GenBank mRNA | AB003587.1 | - | 1 | 221 | ||
| AF334791.1 | - | 1 | 221 | |||
| L48619.1 | - | 2 | 320 | |||
| U75448.1 | - | 1 | 241 | |||

Kcnd3
Genomic Map