siRNA Details
| Gene Symbol: Abcb11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_031760.1 | NP_113948.1 | 5 | 387 | ||
| GenBank mRNA | AF010597.1 | - | 5 | 360 | ||
| U69487.1 | - | 5 | 387 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Abcb11| Gene Symbol: Abcb11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_031760.1 | NP_113948.1 | 5 | 387 | ||
| GenBank mRNA | AF010597.1 | - | 5 | 360 | ||
| U69487.1 | - | 5 | 387 | |||