siRNA Details
| Gene Symbol: Polr1a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_031772.1 | NP_113960.1 | 1, 2 | 133 | ||
| XM_008762998.1 | XP_008761220.1 | 1, 2 | 410 | |||
| GenBank mRNA | AF025425.1 | - | 1, 2 | 133 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Polr1a| Gene Symbol: Polr1a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_031772.1 | NP_113960.1 | 1, 2 | 133 | ||
| XM_008762998.1 | XP_008761220.1 | 1, 2 | 410 | |||
| GenBank mRNA | AF025425.1 | - | 1, 2 | 133 | ||