siRNA Details
| Gene Symbol: Taldo1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_031811.3 | NP_113999.2 | 3 | 253 | ||
| GenBank mRNA | AF069306.1 | - | 3 | 239 | ||
| BC059126.1 | - | 3 | 257 | 6889440 | ||
| FQ213939.1 | - | 3 | 265 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Taldo1| Gene Symbol: Taldo1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_031811.3 | NP_113999.2 | 3 | 253 | ||
| GenBank mRNA | AF069306.1 | - | 3 | 239 | ||
| BC059126.1 | - | 3 | 257 | 6889440 | ||
| FQ213939.1 | - | 3 | 265 | |||