siRNA Details
| Gene Symbol: Rab2a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_031718.1 | NP_113906.1 | 4 | 226 | ||
| GenBank mRNA | FQ229541.1 | - | 4 | 428 | ||
| J02999.1 | - | 4 | 226 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Rab2a| Gene Symbol: Rab2a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_031718.1 | NP_113906.1 | 4 | 226 | ||
| GenBank mRNA | FQ229541.1 | - | 4 | 428 | ||
| J02999.1 | - | 4 | 226 | |||