siRNA Details
| Gene Symbol: Clns1a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_031719.1 | NP_113907.1 | 3 | 345 | ||
| XM_006229792.2 | XP_006229854.1 | 3 | 408 | |||
| GenBank mRNA | BC060555.1 | - | 3 | 389 | 6918578 | |
| D13985.1 | - | 3 | 415 | |||
| D17698.1 | - | 3 | 328 | |||
| FQ212666.1 | - | 1 | 27 | |||
| FQ232880.1 | - | 1 | 27 | |||
| L26450.1 | - | 3 | 345 | |||

Clns1a
Genomic Map