siRNA Details
| Gene Symbol: B4galt6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_031740.1 | NP_113928.1 | 1045 | |||
| GenBank mRNA | AF048687.1 | - | 1045 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

B4galt6| Gene Symbol: B4galt6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_031740.1 | NP_113928.1 | 1045 | |||
| GenBank mRNA | AF048687.1 | - | 1045 | |||