siRNA Details
| Gene Symbol: Slc24a2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_031743.2 | NP_113931.1 | 1 | 248 | ||
| GenBank mRNA | AF021923.1 | - | 1 | 248 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Slc24a2| Gene Symbol: Slc24a2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_031743.2 | NP_113931.1 | 1 | 248 | ||
| GenBank mRNA | AF021923.1 | - | 1 | 248 | ||