siRNA Details
Gene Symbol: Hspb3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031750.1 | NP_113938.1 | 1 | 409 | ||
GenBank mRNA | AF203374.1 | - | 1 | 409 |

More Information
Other Species Targeted


Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Hspb3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031750.1 | NP_113938.1 | 1 | 409 | ||
GenBank mRNA | AF203374.1 | - | 1 | 409 |