siRNA Details
| Gene Symbol: Naaladl1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_031759.1 | NP_113947.1 | 4 | 590 | ||
| GenBank mRNA | AF009921.1 | - | 4 | 590 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Naaladl1| Gene Symbol: Naaladl1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_031759.1 | NP_113947.1 | 4 | 590 | ||
| GenBank mRNA | AF009921.1 | - | 4 | 590 | ||