siRNA Details
| Gene Symbol: TESMIN | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_004923.3 | NP_004914.2 | 7, 8 | 1168 | ||
| XM_011545402.1 | XP_011543704.1 | 7, 8 | 1212 | |||
| XM_011545403.2 | XP_011543705.1 | 6, 7 | 1081 | |||
| XM_017018588.1 | XP_016874077.1 | 5, 6 | 1014 | |||
| GenBank mRNA | BC064579.1 | - | 7, 8 | 1135 | 5743865 | |
| U86074.1 | - | 6, 7 | 807 | |||

TESMIN
Genomic Map