siRNA Details
| Gene Symbol: Galnt5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_031796.1 | NP_113984.1 | 1 | 272 | ||
| GenBank mRNA | AF049344.1 | - | 1 | 272 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Galnt5| Gene Symbol: Galnt5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_031796.1 | NP_113984.1 | 1 | 272 | ||
| GenBank mRNA | AF049344.1 | - | 1 | 272 | ||