siRNA Details
| Gene Symbol: Tpbg | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_031807.1 | NP_113995.1 | 2 | 1252 | ||
| XM_006243499.2 | XP_006243561.1 | 2 | 1542 | |||
| XM_008766457.1 | XP_008764679.1 | 2 | 1353 | |||
| GenBank mRNA | AF063939.1 | - | 2 | 1252 | ||
| BC087011.1 | - | 2 | 1252 | 7193411 | ||

Tpbg
Genomic Map