siRNA Details
| Gene Symbol: Cd164 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_031812.1 | NP_114000.1 | 6 | 521 | ||
| GenBank mRNA | AJ238574.1 | - | 6 | 521 | ||
| BC062064.1 | - | 6 | 524 | 5599253 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Cd164| Gene Symbol: Cd164 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_031812.1 | NP_114000.1 | 6 | 521 | ||
| GenBank mRNA | AJ238574.1 | - | 6 | 521 | ||
| BC062064.1 | - | 6 | 524 | 5599253 | ||