siRNA Details
| Gene Symbol: ROR2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001318204.1 | NP_001305133.1 | 2 | 315 | ||
| NM_004560.3 | NP_004551.2 | 2 | 315 | |||
| XM_005252008.4 | XP_005252065.1 | 2 | 280 | |||
| XM_006717121.3 | XP_006717184.1 | 2 | 937 | |||
| XM_017014762.1 | XP_016870251.1 | 2 | 2012 | |||
| XM_017014763.1 | XP_016870252.1 | 2 | 191 | |||
| GenBank mRNA | AB209154.1 | - | 4 | 584 | ||
| BC033697.1 | - | 2 | 302 | |||
| BC051273.1 | - | 2 | 127 | |||
| BC130522.1 | - | 2 | 203 | 40146553 | ||
| M97639.1 | - | 2 | 315 | |||

ROR2
Genomic Map