siRNA Details
| Gene Symbol: Wisp1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_031716.1 | NP_113904.1 | 5 | 1211 | ||
| GenBank mRNA | AF228049.1 | - | 5 | 1211 | ||
| BC078787.1 | - | 5 | 1193 | 7131232 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Wisp1| Gene Symbol: Wisp1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_031716.1 | NP_113904.1 | 5 | 1211 | ||
| GenBank mRNA | AF228049.1 | - | 5 | 1211 | ||
| BC078787.1 | - | 5 | 1193 | 7131232 | ||