siRNA Details
| Gene Symbol: Dio2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_031720.3 | NP_113908.3 | 2 | 854 | ||
| GenBank mRNA | U53505.1 | - | 2 | 854 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Dio2| Gene Symbol: Dio2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_031720.3 | NP_113908.3 | 2 | 854 | ||
| GenBank mRNA | U53505.1 | - | 2 | 854 | ||