siRNA Details
| Gene Symbol: Htra1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_031721.1 | NP_113909.1 | 3 | 735 | ||
| GenBank mRNA | AF179370.1 | - | 3 | 735 | ||
| BC081767.1 | - | 3 | 760 | 7121959 | ||
| FQ228889.1 | - | 2 | 187 | |||
| FQ228942.1 | - | 2 | 213 | |||

Htra1
Genomic Map