siRNA Details
| Gene Symbol: Scamp4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_031725.1 | NP_113913.1 | 7 | 636 | ||
| XM_006240999.1 | XP_006241061.1 | 7 | 655 | |||
| GenBank mRNA | AF285631.1 | - | 7 | 636 | ||
| BC090342.1 | - | 7 | 630 | 7304207 | ||

Scamp4
Genomic Map