siRNA Details
| Gene Symbol: Aldh3a2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_031731.2 | NP_113919.2 | 6 | 994 | ||
| XM_006246529.2 | XP_006246591.1 | 6 | 1492 | |||
| GenBank mRNA | CB727458.1 | - | 4 | 457 | ||
| FQ210485.1 | - | 6 | 1057 | |||
| FQ221576.1 | - | 6 | 1016 | |||
| M73714.1 | - | 6 | 1000 | |||

Aldh3a2
Genomic Map