siRNA Details
| Gene Symbol: Slc27a2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_031736.1 | NP_113924.1 | 5 | 1901 | ||
| GenBank mRNA | BC081766.1 | - | 5 | 1238 | 7106977 | |
| D85100.1 | - | 5 | 1901 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Slc27a2| Gene Symbol: Slc27a2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_031736.1 | NP_113924.1 | 5 | 1901 | ||
| GenBank mRNA | BC081766.1 | - | 5 | 1238 | 7106977 | |
| D85100.1 | - | 5 | 1901 | |||