siRNA Details
| Gene Symbol: Shank1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_031751.2 | NP_113939.2 | 12, 13 | 1828 | ||
| XM_008759414.1 | XP_008757636.1 | 11, 12 | 1895 | |||
| XM_008759415.1 | XP_008757637.1 | 11, 12 | 1895 | |||
| XM_008759416.1 | XP_008757638.1 | 11, 12 | 1895 | |||
| XM_008759417.1 | XP_008757639.1 | 11, 12 | 1895 | |||
| GenBank mRNA | AF102855.2 | - | 11, 12 | 1740 | ||
| AF131951.1 | - | 11, 12 | 1749 | |||
| AF159046.1 | - | 12, 13 | 1828 | |||

Shank1
Genomic Map