siRNA Details
| Gene Symbol: Cpz | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_031766.1 | NP_113954.1 | 7 | 1166 | ||
| XM_006251151.2 | XP_006251213.1 | 6 | 1161 | |||
| GenBank mRNA | AF017637.1 | - | 7 | 1166 | ||
| AF017638.1 | - | 7 | 1166 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Cpz| Gene Symbol: Cpz | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_031766.1 | NP_113954.1 | 7 | 1166 | ||
| XM_006251151.2 | XP_006251213.1 | 6 | 1161 | |||
| GenBank mRNA | AF017637.1 | - | 7 | 1166 | ||
| AF017638.1 | - | 7 | 1166 | |||