siRNA Details
| Gene Symbol: Pias3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_031784.2 | NP_113972.2 | 5 | 708 | ||
| XM_006232991.2 | XP_006233053.1 | 5 | 961 | |||
| XM_006232992.2 | XP_006233054.1 | 5 | 2043 | |||
| XM_006232993.2 | XP_006233055.1 | 5 | 850 | |||
| XM_008761337.1 | XP_008759559.1 | 5 | 712 | |||
| XM_008761338.1 | XP_008759560.1 | 5 | 961 | |||
| GenBank mRNA | AF032872.2 | - | 5 | 708 | ||

Pias3
Genomic Map