siRNA Details
| Gene Symbol: Trim3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_031786.1 | NP_113974.1 | 7 | 1678 | ||
| GenBank mRNA | AF036255.1 | - | 7 | 1678 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Trim3| Gene Symbol: Trim3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_031786.1 | NP_113974.1 | 7 | 1678 | ||
| GenBank mRNA | AF036255.1 | - | 7 | 1678 | ||