siRNA Details
| Gene Symbol: Nfe2l2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_031789.2 | NP_113977.1 | 5 | 1601 | ||
| XM_006234396.2 | XP_006234458.1 | 5 | 1770 | |||
| XM_006234397.2 | XP_006234459.1 | 5 | 1715 | |||
| XM_006234398.2 | XP_006234460.1 | 5 | 2322 | |||
| GenBank mRNA | AF037350.1 | - | 5 | 1603 | ||
| BC061724.1 | - | 5 | 1637 | 5598313 | ||
| FQ220760.1 | - | 1 | 438 | |||
| FQ221441.1 | - | 3 | 2546 | |||

Nfe2l2
Genomic Map