siRNA Details
| Gene Symbol: Dedd | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_031800.1 | NP_113988.1 | 4 | 692 | ||
| XM_006250269.2 | XP_006250331.1 | 6 | 757 | |||
| XM_006250270.2 | XP_006250332.1 | 4 | 707 | |||
| XM_006250272.2 | XP_006250334.1 | 6 | 837 | |||
| XM_006250273.2 | XP_006250335.1 | 5 | 763 | |||
| XM_006250274.2 | XP_006250336.1 | 4 | 711 | |||
| GenBank mRNA | AF053362.1 | - | 4 | 692 | ||

Dedd
Genomic Map