siRNA Details
| Gene Symbol: Cngb1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_031809.1 | NP_113997.1 | 14 | 1404 | ||
| GenBank mRNA | AJ000496.1 | - | 14 | 1404 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Cngb1| Gene Symbol: Cngb1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_031809.1 | NP_113997.1 | 14 | 1404 | ||
| GenBank mRNA | AJ000496.1 | - | 14 | 1404 | ||