siRNA Details
| Gene Symbol: Wfs1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_031823.1 | NP_114011.1 | 1 | 130 | ||
| GenBank mRNA | AF136378.1 | - | 1 | 130 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Wfs1| Gene Symbol: Wfs1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_031823.1 | NP_114011.1 | 1 | 130 | ||
| GenBank mRNA | AF136378.1 | - | 1 | 130 | ||