siRNA Details
| Gene Symbol: Flot2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001270801.1 | NP_001257730.1 | 2 | 173 | ||
| NM_031830.2 | NP_114018.1 | 3 | 255 | |||
| GenBank mRNA | AF023303.1 | - | 3 | 234 | ||
| AF023304.1 | - | 2 | 152 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Flot2| Gene Symbol: Flot2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001270801.1 | NP_001257730.1 | 2 | 173 | ||
| NM_031830.2 | NP_114018.1 | 3 | 255 | |||
| GenBank mRNA | AF023303.1 | - | 3 | 234 | ||
| AF023304.1 | - | 2 | 152 | |||