siRNA Details
| Gene Symbol: Trpv1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_031982.1 | NP_114188.1 | 3 | 432 | ||
| XM_008767882.1 | XP_008766104.1 | 2 | 439 | |||
| GenBank mRNA | AB040873.1 | - | 2 | 352 | ||
| AB041029.1 | - | 2 | 352 | |||
| AF029310.1 | - | 3 | 432 | |||

Trpv1
Genomic Map