siRNA Details
| Gene Symbol: Eif2b2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_032058.2 | NP_114447.2 | 2, 3 | 323 | ||
| GenBank mRNA | BC070506.1 | - | 2, 3 | 323 | 7097686 | |
| FQ232990.1 | - | 2, 3 | 332 | |||
| U31880.1 | - | 2, 3 | 326 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Eif2b2| Gene Symbol: Eif2b2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_032058.2 | NP_114447.2 | 2, 3 | 323 | ||
| GenBank mRNA | BC070506.1 | - | 2, 3 | 323 | 7097686 | |
| FQ232990.1 | - | 2, 3 | 332 | |||
| U31880.1 | - | 2, 3 | 326 | |||