siRNA Details
| Gene Symbol: Kalrn | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_032062.2 | NP_114451.2 | 128 | |||
| GenBank mRNA | AF230644.1 | - | 128 | |||
| AF232668.1 | - | 98 | ||||
| AF232669.1 | - | 98 | ||||
| AY621095.1 | - | 98 | ||||
| U88156.1 | - | 481 | ||||
| U88157.1 | - | 98 | ||||

Kalrn
Genomic Map