siRNA Details
| Gene Symbol: Hsd17b12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_032066.1 | NP_114455.1 | 2 | 186 | ||
| XM_006234599.2 | XP_006234661.1 | 2 | 353 | |||
| GenBank mRNA | BC061543.1 | - | 2 | 207 | 6888949 | |
| FQ213557.1 | - | 2 | 222 | |||
| FQ214777.1 | - | 2 | 251 | |||
| U81186.1 | - | 2 | 186 | |||

Hsd17b12
Genomic Map