siRNA Details
| Gene Symbol: Kcnq1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_032073.1 | NP_114462.1 | 4 | 752 | ||
| GenBank mRNA | AJ133685.1 | - | 4 | 752 | ||
| AY043394.1 | - | 3 | 492 | |||
| U92655.1 | - | 3 | 264 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Kcnq1| Gene Symbol: Kcnq1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_032073.1 | NP_114462.1 | 4 | 752 | ||
| GenBank mRNA | AJ133685.1 | - | 4 | 752 | ||
| AY043394.1 | - | 3 | 492 | |||
| U92655.1 | - | 3 | 264 | |||