siRNA Details
| Gene Symbol: Rab11b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_032617.2 | NP_116006.1 | 2 | 241 | ||
| GenBank mRNA | AF286534.1 | - | 2 | 247 | ||
| BC062041.1 | - | 2 | 241 | 5598168 | ||
| D01046.1 | - | 2 | 227 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Rab11b| Gene Symbol: Rab11b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_032617.2 | NP_116006.1 | 2 | 241 | ||
| GenBank mRNA | AF286534.1 | - | 2 | 247 | ||
| BC062041.1 | - | 2 | 241 | 5598168 | ||
| D01046.1 | - | 2 | 227 | |||