siRNA Details
| Gene Symbol: Sec31a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_033021.1 | NP_148981.1 | 3, 4 | 472 | ||
| GenBank mRNA | AF034582.1 | - | 3, 4 | 472 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Sec31a| Gene Symbol: Sec31a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_033021.1 | NP_148981.1 | 3, 4 | 472 | ||
| GenBank mRNA | AF034582.1 | - | 3, 4 | 472 | ||